Cole-Parmer® Spatula Macro Spoons, Stainless Steel
Polished surface provides fast and easy cleaning
  • Chemical and corrosion resistant
  • Available in 6 sizes
The AISI 304 grade stainless steel spatula/macro spoon combinations are chemical and corrosion resistant. One end features a macro spoon for medium and large size particles, while the other end offers a straight/flat spatula. Robust and seamless construction guarantees long life cycle.
1,900.00 - ₹5,500.00INR / Pkg of 3
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Overall Length (in) Width (in)
Availability Pricing
COMPARE
304 stainless steel 4 45/64 29/32
₹1,900.00INR / Pkg of 3
COMPARE
304 stainless steel 5 29/32 1 1/10
₹2,200.00INR / Pkg of 3
COMPARE
304 stainless steel 7 1/10 1 1/5
₹2,500.00INR / Pkg of 3
COMPARE
304 stainless steel 8 19/64 1 1/5
₹2,900.00INR / Pkg of 3
COMPARE
304 stainless steel 9 51/64 1 1/2
₹4,100.00INR / Pkg of 3
COMPARE
304 stainless steel 11 51/64 1 45/64
₹5,500.00INR / Pkg of 3