Cole-Parmer Spatula Macro Spoon, Stainless Steel,180mm; 3/PK
Representative image only.
Cole-Parmer Item # 0628706
Polished surface provides fast and easy cleaning
  • Chemical and corrosion resistant
  • Available in 6 sizes
PRODUCT OPTIONS
₹2,500.00INR / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)7 1/10
  • Width (in)1 1/5
  • Overall Length (cm)18
  • Width (cm)3.1
  • Material304 stainless steel
  • ColorMetalic
More About this Item
The AISI 304 grade stainless steel spatula/macro spoon combinations are chemical and corrosion resistant. One end features a macro spoon for medium and large size particles, while the other end offers a straight/flat spatula. Robust and seamless construction guarantees long life cycle.