Cole-Parmer Spatula/Micro Spoon, Stainless Steel,150mm; 3/PK
Representative image only.
Cole-Parmer Item # 0628716
Easily handles small samples
  • Chemical and corrosion resistant
  • Easy to clean
PRODUCT OPTIONS
₹2,100.00INR / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)5 29/32
  • Overall Length (cm)15
  • Material304 stainless steel
  • ColorMetalic
  • DescriptionSpatula Micro Spoon, SS,150mm; 3/PK
Key Features
  • Chemical and corrosion resistant
  • Easy to clean
More About this Item
The AISI 304 grade stainless steel spatula/micro spoon combinations are chemical and corrosion resistant. They are ideal for handling small size objects.
Features a micro spoon for small size particles at one end and a straight/flat spatula at the other end. Robust and seamless construction guarantees long life cycle. Smooth polished surfaces provide fast and easy cleaning.