Cole-Parmer® Spatulas/Micro Spoons, SS
Easily handles small samples
  • Chemical and corrosion resistant
  • Easy to clean
The AISI 304 grade stainless steel spatula/micro spoon combinations are chemical and corrosion resistant. They are ideal for handling small size objects.
Features a micro spoon for small size particles at one end and a straight/flat spatula at the other end. Robust and seamless construction guarantees long life cycle. Smooth polished surfaces provide fast and easy cleaning.
2,100.00 - ₹2,300.00INR / Pkg of 3
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Overall Length (cm) Overall Length (in)
Availability Pricing
COMPARE
304 stainless steel 15 5 29/32
₹2,100.00INR / Pkg of 3
COMPARE
304 stainless steel 18 7 3/32
₹2,300.00INR / Pkg of 3
COMPARE
304 stainless steel 21 8 17/64
₹2,300.00INR / Pkg of 3