CPC (Colder) AseptiQuik W® Quick-Disconnect Fitting, High-Flow Large-Bore, Genderless Sanitary, High-Temperature Polycarbonate, 1-1/2" Sanitary; 1/Ea

Representative image only.
CPC (Colder) Mfr # AQW33024HT – Item # 5011213
High-flow, large-bore genderless sterile connectors simplify process integration while maximizing flexibility
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, pull, and snap” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Ideal for large-volume, high-flow applications
  • Robust construction provides reliable and repeatable fluid transfer
  • Sterilize by autoclave up to 130°C (266°F)
  • 0.25" nominal flow path size
  • 1-1/2" sanitary connection

₹45,800.00INR / Each
Often Bought With

SPECS & DESCRIPTION

Specifications & Description

  • Fitting MaterialPolycarbonate
  • Seal MaterialPlatinum-Cured Silicone
  • Sterilization MethodGamma irradiation up to 50 kGy
  • Connection TypeQuick Disconnect x Sanitary
  • Sanitary Clamp Size (Port 1) (in)1-1/2
  • Flow PatternStraight
  • ColorWhite with white pull tab, gray pull tab guide, and blue side latch
  • Min Temperature (° F)34
  • Max Temperature (° F)104
  • Min Temperature (° C)1
  • Max Temperature (° C)40
  • Max Operating Pressure (PSI)60
  • Fitting Length (in)3 11/16
  • Fitting Width (in)5 3/4
  • Fitting Length (mm)93
  • Fitting Width (mm)146.6
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;FDA;RoHS;USP;ADCF
  • DescriptionQuick-Disconnect Fitting, High-Flow Large-Bore, Genderless Sanitary, High-Temperature Polycarbonate, 1-1/2" Sanitary; 1/Ea
Certificate of Compliance

Key Features

  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, pull, and snap” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Ideal for large-volume, high-flow applications
  • Robust construction provides reliable and repeatable fluid transfer
  • Sterilize by autoclave up to 130°C (266°F)
  • 0.25" nominal flow path size
  • 1-1/2" sanitary connection

More About this Item

The AseptiQuik® genderless connectors provide fast, easy, secure connections and quick assembly. The interior of the flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, simply unsnap and flip down the protective tab cover, align connectors with the pull tabs hanging down, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, pull the tabs to simultaneously remove both membranes from the connector halves, remove shipping clips, then snap on each side latch.

All AseptiQuik connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot. High-temperature polycarbonate withstands autoclaving up to 130°C (266°F) for 60 minutes.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Certificate of Compliance

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat