CPC (Colder) EFC12 Quick-Disconnect Fittings, Hose Barb Body, Polypropylene, Valved, 1/4" ID

Representative image only.
CPC (Colder) Mfr # EFCD17412 – Item # 5013520
High efficiency valve provides more flow in a compact size
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Chemically resistant and gamma sterilizable
  • Ideal for tubing sizes from 1/4“ to 3/8” ID
  • Compatible with most APC couplings
  • 1/4" hose barb

PRODUCT OPTIONS

₹4,100.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Fitting MaterialPolypropylene
  • Valve MaterialPolypropylene
  • Valve Spring MaterialStainless steel
  • O-Ring MaterialEPDM
  • Connection TypeQuick-Disconnect Hose Barb Body, Valve
  • Barb Size / Tubing Id (Port 1) (in)1/4
  • Flow PatternStraight
  • Fitting Length (in)2 15/64
  • Fitting Width (in)59/64
  • Fitting Length (mm)56.6
  • Fitting Width (mm)23.6

Key Features

  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Chemically resistant and gamma sterilizable
  • Ideal for tubing sizes from 1/4“ to 3/8” ID
  • Compatible with most APC couplings
  • 1/4" hose barb

More About this Item

The CPC (Colder) EFC Series quick-disconnect fittings offer a high-efficiency valve design, delivering superior flow capacity in a compact size. Featuring a plastic thumb latch with no exposed metal, these fittings ensure easy, gloved-hand operation. Made from chemically resistant polypropylene, they are ideal for harsh environments, providing durability and reliable performance.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient-oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat