CPC (Colder) MPC™ Quick-Disconnect Fitting, Cap with Lock, Polysulfone; 25/Pk

Representative image only.
CPC (Colder) Mfr # MPCK32039B25 – Item # 5011300
Connect to a male coupling insert to stop the flow
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Operate ergonomic thumb latch even with gloved hands
  • Locking sleeve prevents accidental disconnection
  • Polysulfone; 25/Pk

PRODUCT OPTIONS

₹34,600.00INR / Pkg of 25
Use caps with coupling inserts

SPECS & DESCRIPTION

Specifications & Description

  • Fitting MaterialPolysulfone
  • Locking Sleeve MaterialPolysulfone
  • Sterilization MethodGamma irradiation up to 50 kGy or autoclave up to 132°C (270°F) for 60 minutes, up to 25 repetitions
  • Connection TypeCap
  • Number Of Ports1
  • Flow PatternCap
  • ColorAmber tint
  • Min Temperature (° F)-40
  • Max Temperature (° F)300
  • Min Temperature (° C)-40
  • Max Temperature (° C)149
  • Max Operating Pressure (PSI)60
  • Fitting Length (in)5/16
  • Fitting Width (in)1
  • Fitting Length (mm)33
  • Fitting Width (mm)25.2
  • Regulatory ApprovalsNon-DEHP;ADCF;RoHS;FDA;Non-Phthalate;USP
  • DescriptionQuick-Disconnect Fitting, Cap with Lock, Polysulfone; 25/Pk

Key Features

  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Operate ergonomic thumb latch even with gloved hands
  • Locking sleeve prevents accidental disconnection
  • Polysulfone; 25/Pk

More About this Item

The CPC® (Colder) MPC™ series connectors provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. Use a cap with an insert to seal off the fluid pathway. Order connectors with a locking sleeve to keep two halves together to guard against accidental disconnection.

All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Certificate of Compliance

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat