CPC (Colder) MPU® Quick-Disconnect Fitting, Twist-to-Connect, Polysulfone, Cap; 1/Ea
Representative image only.
CPC (Colder) Mfr # MPU32039 – Item # 5011306
Connect to a male coupling insert to stop the flow
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Easy-to-use twist-to-connect design with locking mechanism
₹2,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolysulfone
  • Sterilization MethodGamma irradiation up to 50 kGy or autoclave up to 132°C (270°F) for 60 minutes, up to 25 repetitions
  • Number Of Ports1
  • Connection TypeCap
  • Flow PatternCap
  • ColorAmber tint
  • Min Temperature (° F)-40
  • Fitting Length (in)25/32
  • Fitting Width (in)1 3/4
  • Fitting Length (mm)20.1
  • Fitting Width (mm)44.5
  • Regulatory ApprovalsRoHS;FDA;Non-DEHP;Non-Phthalate;USP;ADCF
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Easy-to-use twist-to-connect design with locking mechanism
More About this Item
The CPC® (Colder) MPU® series connectors provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. The special twist-to-connect design features a locking mechanism that ensures against accidental disconnects. Use a cap with an insert to seal off the fluid pathway.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.