CPC (Colder) NS4 Quick-Disconnect Fittings, Threaded Body, Polypropylene, Valved, 3/8" PTF

Representative image only.
CPC (Colder) Mfr # NS4D13006 – Item # 5013590
Designed for a pressure-activated release without leaks
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Chemically resistant
  • Ideal for tubing sizes from 1/8” to 3/8” ID
  • Designed for semi-rigid tubing
  • 3/8" PTF

₹5,300.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Fitting MaterialPolypropylene
  • Valve Spring Material316 Stainless steel
  • O-Ring MaterialEPDM
  • Chemical ResistanceYes
  • Connection TypeQuick-Disconnect Threaded Body, Valve
  • Thread Size NPT(F) (Port 1) (InchShort)3/8
  • Flow PatternStraight
  • ColorGray with dark gray overmold standard
  • Min Temperature (° F)32
  • Max Temperature (° F)160
  • Min Temperature (° C)0
  • Max Temperature (° C)71
  • Max Operating Pressure (PSI)120
  • Fitting Length (in)2 29/64
  • Fitting Width (in)1 1/20
  • Fitting Length (mm)62.2
  • Fitting Width (mm)26.7

Key Features

  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Chemically resistant
  • Ideal for tubing sizes from 1/8” to 3/8” ID
  • Designed for semi-rigid tubing
  • 3/8" PTF

More About this Item

The CPC (Colder) NS4 Series quick-disconnect threaded insert fitting is designed with non-spill valves to prevent spills, reduce downtime, and improve operator safety. This lightweight, compact fitting offers excellent chemical resistance and user-friendly operation.

An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. The fitting operates efficiently in temperatures ranging from 32 to 160 °F (0 to 71 °C) and withstands pressures from vacuum to 120 psi (8.3 bar).

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient-oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat