Digi-Sense Data Acquisition Software with USB Cable; Cable 2
Representative image only.
Item # 3780417
Capture and send data right to your computer
  • Compatible with any PC with Windows®
  • 1 year warranty
PRODUCT OPTIONS
₹27,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With37950-03, 37950-11, 19601-03, 37950-02
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Compatible with any PC with Windows®
  • 1 year warranty
More About this Item
Complete Data Acquisition Software enables you to display and store measurement readings on your PC from up to 8 devices at a time. Data capture runs in the background, enabling you to work on other programs, simultaneously. Set audible and visual notification alarms or immediately send an email if a over/under condition occurs. The software can be used to connect a meter directly to your PC via cable, or coupled with the Digi-Sense Wireless Radio-Signal Transceiver Kit (37804-12). Check software specification tables for meter compatibility, or contact our application specialists if you have a meter with RS-232 output not listed in the tables.