Traceable® Data Acquisition Software
Capture and send data right to your computer
  • Compatible with any PC with Windows®
Complete Data Acquisition Software enables you to display and store measurement readings on your PC from up to 8 devices at a time. Data capture runs in the background, enabling you to work on other programs, simultaneously. Set audible and visual notification alarms or immediately send an email if a over/under condition occurs. The software can be used to connect a meter directly to your PC via cable, or coupled with the Digi-Sense Wireless Radio-Signal Transceiver Kit (37804-12). Check software specification tables for meter compatibility, or contact our application specialists if you have a meter with RS-232 output not listed in the tables.
25,000.00 - ₹64,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Data Acquisition Software with USB Cable; Cable 1
₹25,000.00INR / Each
COMPARE
Data Acquisition Software with USB Cable; Cable 2
₹27,600.00INR / Each
COMPARE
Data Acquisition Software with USB Cable and Calibration; Cable 3
₹64,300.00INR / Each