DWK Life Sciences (Kimble) HPLC Reservoir System, Bottle/Cap/Sparge/Tubing; 5.0 Liter
Representative image only.
DWK Life Sciences (Kimble) Mfr # 953930-5002 – Item # 0201755
Coated glass bottles prevent photodegradation of light-sensitive mobile phases
  • Constructed of durable safety coated glass
  • 1 year warranty
PRODUCT OPTIONS
₹205,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • End CapEconomy
  • Bottom TypeFlat
  • Total Volume Capacity (mL)5000
  • DescriptionHPLC Reservoir System, Bottle/Cap/Sparge/Tubing; 5.0 Liter
  • Warranty1 year
1 Year Warranty
Key Features
  • Constructed of durable safety coated glass
  • 1 year warranty
More About this Item
These complete reservoir systems have chemical-resistant fluid / vapor contact surfaces. The cap body is PTFE with a TFE/propylene o-ring and a polypropylene screw collar. Tubing passes through ¨ù-28 fittings. Cap has two 1/8" through holes and one through hole for 1/16¡± OD tubing. Includes one 2 µm PEEK Inlet/Sparge filter, two ETFE fittings, one ETFE plug, and 10 ft of 1/8" OD FEP PTFE tubing.
The ULTRA-WARE Flat Bottom Reservoir is Type I, Class A borosilicate glass with a GL 45-4 screw thread and a plastic safety coating. The plastic coating blocks UV light up to 385 nm.