Kimble® ULTRA-WARE® Economy Cap Systems
Coated glass bottles prevent photodegradation of light-sensitive mobile phases
  • Constructed of durable safety coated glass
These complete reservoir systems have chemical-resistant fluid / vapor contact surfaces. The cap body is PTFE with a TFE/propylene o-ring and a polypropylene screw collar. Tubing passes through ¨ù-28 fittings. Cap has two 1/8" through holes and one through hole for 1/16¡± OD tubing. Includes one 2 µm PEEK Inlet/Sparge filter, two ETFE fittings, one ETFE plug, and 10 ft of 1/8" OD FEP PTFE tubing.
The ULTRA-WARE Flat Bottom Reservoir is Type I, Class A borosilicate glass with a GL 45-4 screw thread and a plastic safety coating. The plastic coating blocks UV light up to 385 nm.
107,000.00 - ₹205,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Bottom Type Description
Availability Pricing
DWK Life Sciences (Kimble) HPLC Reservoir System, Bottle/Cap/Sparge/Tubing; 1.0 Liter 0201745 Mfr # 953930-1002
COMPARE
1000 Flat
₹107,000.00INR / Each
DWK Life Sciences (Kimble) HPLC Reservoir System, Bottle/Cap/Sparge/Tubing; 2.0 Liter 0201750 Mfr # 953930-2002
COMPARE
2000 Flat
₹120,000.00INR / Each
DWK Life Sciences (Kimble) HPLC Reservoir System, Bottle/Cap/Sparge/Tubing; 5.0 Liter 0201755 Mfr # 953930-5002
COMPARE
5000 Flat
₹205,000.00INR / Each