DWK Life Sciences (Kimble) Silicone Lid, Large; Cyan
Representative image only.
DWK Life Sciences (Kimble) Mfr # 291113129 – Item # 3450025
Versatile lids work on various vessel diameters and with spouts, beads, or thread finishes
  • Constructed of stretchable, chemically resistant, and heat-resistant silicone
  • Quickly identify or track samples visually with different color options
  • Reusable dishwasher-safe lids reduce environmental impact and lowers costs
  • Large lid has a 84 to 116 mm diameter
₹880.00INR / Each
OFTEN BOUGHT WITH…
KIMAX® Color-Coded Glass Griffin Beakers
14,200.00 - ₹28,300.00INR / Case of 12
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSilicone
  • Cap MaterialSilicone
  • Cap / Stopper SizeLarge
  • Cap ColorCyan
  • Special Features84 to 116 mm diameter
  • Withstands Low Temperature (° F)-40
  • AutoclavableYes
  • Withstands High Temperature (° F)346
Key Features
  • Constructed of stretchable, chemically resistant, and heat-resistant silicone
  • Quickly identify or track samples visually with different color options
  • Reusable dishwasher-safe lids reduce environmental impact and lowers costs
  • Large lid has a 84 to 116 mm diameter
More About this Item
These DWK KIMBLE® silicone lids are made of stretchable silicone to fit on beakers, flasks, bottles, and cylinders of various diameter vessels. The stretchable yet secure lids provide a tight fit on vessels with spouts, beads, and threaded finishes. This versatility allows for many uses beyond the limits of traditional films and covers. Lids are available in three colors for quick visual identification—pink, cyan, and green. The stretchable silicone material is compatible with lab markers to allow for optimal sample identification.