Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/8" NPT(M) x 1/8" ID
Representative image only.
Eldon James Mfr # A2-2SS – Item # 5012459
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
  • 1/8" NPT(M) x 1/8" hose barb
PRODUCT OPTIONS
₹4,300.00INR / Each
Often Bought With
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless Steel
  • Sterilization MethodE-beam/gamma irradiation, EtO, or autoclave up to 134°C
  • Connection TypeThreaded NPT(M) x Hose Barb
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)1/8
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Flow PatternStraight
  • ColorMetallic
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;RoHS
  • DescriptionThreaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/8" NPT(M) x 1/8" ID
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
  • 1/8" NPT(M) x 1/8" hose barb
More About this Item
Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place.
These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.