EMD Millipore UFC501024 Centrifugal Filter Units, 10k NMWL, 24/pk, green

Representative image only.
EMD Millipore Mfr # UFC501024 – Item # 2995050
Highest recovery of sample with solute recoveries typically >95%
  • Membrane materials: Ultracel-YM regenerated cellulose
  • Membrane support base: acetal
  • Sample reservoir: polypropylene
  • O-ring: medical-grade silicone rubber
  • Sample vial: polycarbonate
  • 1 year warranty

PRODUCT OPTIONS

₹75,100.00INR / Pkg of 24

SPECS & DESCRIPTION

Specifications & Description

  • ColorGreen
  • MembraneUltracel-um Regenerated Cellulose
  • Nominal Molecular Weight Limit (Mwl) (Dalton)10000
  • DescriptionCentrifugal Filter Units, 10k NMWL, 24/pk, green
  • Warranty1 year
1 Year Warranty

Key Features

  • Membrane materials: Ultracel-YM regenerated cellulose
  • Membrane support base: acetal
  • Sample reservoir: polypropylene
  • O-ring: medical-grade silicone rubber
  • Sample vial: polycarbonate
  • 1 year warranty

More About this Item

Ideal for dilute (ng/mL to µg/mL range) protein solutions. Low-binding ultracel-YM regenerated cellulose membrane available with different molecular weight cut-offs. Used in standard 1.5 mL tube fixed-angle rotors. Applications include concentrating and purifying proteins, antibodies and nucleic acids (alternative to EtOH precipitation), desalting and buffer exchange, and removal of primers, linkers and unin-corporated label.

Maximum initial sample volume is 500 µl, and typical final concentrate volume is 5 to 15 µl. Maximum centrifugal force is 14,000g , 500g for 29950-54 on DNA/RNA applications. Maximum initial sample volume is 500 ul, and typical final concentrate volume is 5 to 15 ul. Maximum centrifugal force is 14,000g , 500g for 29950-54 on DNA/RNA applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat