Millipore® Amicon® Centrifugal Filter Units

Highest recovery of sample with solute recoveries typically >95%
  • Membrane materials: Ultracel-YM regenerated cellulose
  • Membrane support base: acetal
  • Sample reservoir: polypropylene
  • O-ring: medical-grade silicone rubber
  • Sample vial: polycarbonate
Ideal for dilute (ng/mL to µg/mL range) protein solutions. Low-binding ultracel-YM regenerated cellulose membrane available with different molecular weight cut-offs. Used in standard 1.5 mL tube fixed-angle rotors. Applications include concentrating and purifying proteins, antibodies and nucleic acids (alternative to EtOH precipitation), desalting and buffer exchange, and removal of primers, linkers and unin-corporated label.

Maximum initial sample volume is 500 µl, and typical final concentrate volume is 5 to 15 µl. Maximum centrifugal force is 14,000g , 500g for 29950-54 on DNA/RNA applications. Maximum initial sample volume is 500 ul, and typical final concentrate volume is 5 to 15 ul. Maximum centrifugal force is 14,000g , 500g for 29950-54 on DNA/RNA applications.

75,100.00INR / Pkg of 24

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Nominal Molecular Weight Limit (Mwl) (Dalton)
Availability Pricing
EMD Millipore UFC501024 Centrifugal Filter Units, 10k NMWL, 24/pk, green 2995050 Mfr # UFC501024
COMPARE
10000
₹75,100.00INR / Pkg of 24
EMD Millipore 42412 Centrifugal Filter Units, 100k NMWL, 24/pk, blue 2995054 Mfr # UFC510024
COMPARE
100000
₹75,100.00INR / Pkg of 24
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat