Environmental Express Environmental Express/ Oakton® Autoclavable pH Probe, Sealed/DJ/Glass; 210 mm
Representative image only.
Environmental Express Item # 0599396
Sealed electrodes improve reading stability
  • Sealed, gel-filled, double-junction reference electrodes require no refilling
  • Offers long-term stability
  • Extends service life
  • 1 year warranty
PRODUCT OPTIONS
₹61,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsAutoclavable, sealed
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Max Temperature (° C)130
  • ConnectorDepends upon cable ordered separately: BNC or US standard
  • Warranty1 year
1 Year Warranty
Key Features
  • Sealed, gel-filled, double-junction reference electrodes require no refilling
  • Offers long-term stability
  • Extends service life
  • 1 year warranty
More About this Item
These electrodes are designed for insertion in RADIOMETER mountings or Ingold nonpressurized assemblies 761-351, 761-361, and 361CH-361 (12 mm diameter)
Length listed in table refers to probe immersion length, which is equal to housing length plus 70 mm (except Ingold 761-31, where immersion length equals housing length plus 50 mm).