Environmental Express FlipMate SC0306C Filtration Systems, Acid Wash Glass, >10 um, Green, 100 mL; 100/Pk
Representative image only.
Environmental Express Item # 3520226
Convenient, economical method for filtration
  • Produce quality results with clean samples
  • Results within seconds
  • 3 month warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Pore Size (µm)10
  • Body Material Acid Wash Glass; green caps
  • Funnel Size (mL)100
  • DescriptionFiltration Systems, Acid Wash Glass, >10 um, Green, 100 mL; 100/Pk
  • Warranty3 month
3 Month Warranty
Key Features
  • Produce quality results with clean samples
  • Results within seconds
  • 3 month warranty
More About this Item
The FlipMate system easily filters the sample with the use of two threaded digestion cups, a filter assembly, vacuum tubing, and vacuum pressure source. The sample is placed in one cup, the filter assembly threaded onto the top of the cup, then the cup and filter are flipped, the receiving cup attached, vaccum tubing attached, vaccum pressure applied, and the sample is then pulled from the sample cup, through the filter assembly, and into the empty cup.
Systems include digestion cups and filtration assemblies. Tubing and vacuum source are not included.