Environmental Express® FlipMate Filtration Systems
Convenient, economical method for filtration
  • Produce quality results with clean samples
  • Results within seconds
The FlipMate system easily filters the sample with the use of two threaded digestion cups, a filter assembly, vacuum tubing, and vacuum pressure source. The sample is placed in one cup, the filter assembly threaded onto the top of the cup, then the cup and filter are flipped, the receiving cup attached, vaccum tubing attached, vaccum pressure applied, and the sample is then pulled from the sample cup, through the filter assembly, and into the empty cup.
Systems include digestion cups and filtration assemblies. Tubing and vacuum source are not included.
50,300.00 - ₹83,400.00INR / Pkg of 100
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Funnel Size (mL) Pore Size (µm)
Availability Pricing
COMPARE
PES with PTFE pre-filter; green caps 50 0.45
₹62,800.00INR / Pkg of 200
COMPARE
PES with PTFE pre-filter; white caps 50 0.45
₹63,600.00INR / Pkg of 200
COMPARE
Glass; green caps 50 1.5
₹61,400.00INR / Pkg of 200
COMPARE
Glass; white caps 50 1.5
₹62,600.00INR / Pkg of 200
COMPARE
PES; green caps 50 0.45
₹61,500.00INR / Pkg of 200
COMPARE
Certified; PES with PTFE pre-filter; green caps 50 0.45
₹83,400.00INR / Pkg of 200
COMPARE
Certified; glass with PTFE pre-filter; green caps 50 0.7
₹63,700.00INR / Pkg of 200
COMPARE
PES with pre-filter 100 0.45
₹50,300.00INR / Pkg of 100
COMPARE
Glass; green caps 100 1.5
₹51,000.00INR / Pkg of 100
COMPARE
PES with PTFE pre-filter; green caps 100 0.2
₹51,700.00INR / Pkg of 100
COMPARE
Acid-washed glass; green caps 100 0.7
₹51,000.00INR / Pkg of 100
COMPARE
Acid Wash Glass; green caps 100 10
Call For Price
COMPARE
Certified; glass with PTFE pre-filter; green caps 100 0.45
₹67,600.00INR / Pkg of 100
COMPARE
Polypropylene, white caps 50 0.7
₹62,800.00INR / Pkg of 200
Showing 1- 14 of 14