Environmental Express Pump, Linear, IAQ Sampling; 120V

Representative image only.
Environmental Express Item # 6006207
Compact yet powerful—produces a high volume of vacuum airflow
  • Produces flow rates up to 30 L/min
  • Comes with flowmeter and regulator valve
  • Ideal for use with Air-O-Cell® cassettes
  • 120 VAC operation
  • 1 year warranty

PRODUCT OPTIONS

₹57,800.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Power (VAC)120
  • DescriptionPump, Linear, IAQ Sampling; 120 VAC
  • Warranty1 year
1 Year Warranty

Includes

Flowmeter, regulator, tubing, on/off switch, and power cord with US standard plug.

Key Features

  • Produces flow rates up to 30 L/min
  • Comes with flowmeter and regulator valve
  • Ideal for use with Air-O-Cell® cassettes
  • 120 VAC operation
  • 1 year warranty

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat