Zefon Z-Lite™ IAQ Sampling Pump
Compact yet powerful—produces a high volume of vacuum airflow
  • Produces flow rates up to 30 L/min
  • Comes with flowmeter and regulator valve
  • Ideal for use with Air-O-Cell® cassettes
57,800.00 - ₹60,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
120
₹57,800.00INR / Each
COMPARE
230
₹60,600.00INR / Each