Environmental Express Zefon® Starter Kit, Basic Mold Sampling; 230V
Representative image only.
Environmental Express Item # 6006204
Quickly and easily sample for mold
  • Convenient kits include everything you need to check for presence of mold
  • Basic kit for air sampling for mold
  • 230 VAC operation
  • 1 year warranty
PRODUCT OPTIONS
₹109,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Flow Rate (LPM)30
  • Included Accessories10 Air-O-Cell® cassettes
  • Power (VAC)230
  • DescriptionStarter Kit, Basic Mold Sampling; 230 VAC
  • Warranty1 year
1 Year Warranty
Includes
IAQ air sampling pump with flowmeter/regulator valve and power cord with European plug, tubing, stand, bioisolation filter, carrying case, and 10 Air-O-Cell® cassettes.
Key Features
  • Convenient kits include everything you need to check for presence of mold
  • Basic kit for air sampling for mold
  • 230 VAC operation
  • 1 year warranty
More About this Item
Each starter kit includes a Z-Lite™ IAQ sampling pump with flowmeter/regulator valve and power cord with plug (US standard for 120 VAC; European for 230 VAC), tubing, stand, bioisolation filter, carrying case, plus sampling media. Select from basic, deluxe, or ultimate kit depending on your sampling needs.
Simply use the included tubing to connect the Air-O-Cell® cassettes to the Z-Lite IAQ sampling pump to collect air samples. Bio-Tape™ slides and swabs (deluxe and ultimate kits only) make it convenient to take surface samples. Connect inner wall sampling adapter (ultimate kit only) to Air-O-Cell cassette to collect samples from inside the wall. Once the sampling is complete, send the samples to a laboratory for analysis.
Included media have a limited shelf life.