Zefon Mold Sampling Starter Kits
Quickly and easily sample for mold
  • Convenient kits include everything you need to check for presence of mold
Each starter kit includes a Z-Lite™ IAQ sampling pump with flowmeter/regulator valve and power cord with plug (US standard for 120 VAC; European for 230 VAC), tubing, stand, bioisolation filter, carrying case, plus sampling media. Select from basic, deluxe, or ultimate kit depending on your sampling needs.
Simply use the included tubing to connect the Air-O-Cell® cassettes to the Z-Lite IAQ sampling pump to collect air samples. Bio-Tape™ slides and swabs (deluxe and ultimate kits only) make it convenient to take surface samples. Connect inner wall sampling adapter (ultimate kit only) to Air-O-Cell cassette to collect samples from inside the wall. Once the sampling is complete, send the samples to a laboratory for analysis.
100,000.00 - ₹153,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Starter Kit, Basic Mold Sampling; 120 VAC
₹100,000.00INR / Each
COMPARE
Starter Kit, Basic Mold Sampling; 230 VAC
₹109,000.00INR / Each
COMPARE
Starter Kit, Deluxe Mold Sampling; 120 VAC
₹116,000.00INR / Each
COMPARE
Starter Kit, Deluxe Mold Sampling; 230 VAC
₹120,000.00INR / Each
COMPARE
Starter Kit, Ultimate Mold Sampling; 120 VAC
₹140,000.00INR / Each
COMPARE
Starter Kit, Ultimate Mold Sampling; 230 VAC
₹153,000.00INR / Each