Eppendorf epTIPS Single Use Racks 50 to 1000 uL 480/Pk

Representative image only.
Eppendorf Mfr # 0030075250 – Item # 1888812
High standards of purity ideally suited for critical laboratory applications
  • Pyrogen-free (endotoxin-free) according to LAL test
  • Sterilized by beta radiation to SAL 10-6, avoiding contamination by ethylene oxide
  • Tested and certified by an independent laboratory
  • Choose Biopur tips for molecular and microbiology, cell technology, quality control, and forensic applications

PRODUCT OPTIONS

₹36,500.00INR / Case of 480

SPECS & DESCRIPTION

Specifications & Description

  • SterileNo
  • Max Volume (µL)1000
  • Min Volume (µL)50
  • Min Volume (mL)0.05
  • Max Volume (mL)1
  • Tip Volume (mL)1
  • Min Dispensing (mL)0.05
  • Max Dispensing (mL)1
  • Package OfIndividually Wrapped Single Use Racks; 480/Pk

Key Features

  • Pyrogen-free (endotoxin-free) according to LAL test
  • Sterilized by beta radiation to SAL 10-6, avoiding contamination by ethylene oxide
  • Tested and certified by an independent laboratory
  • Choose Biopur tips for molecular and microbiology, cell technology, quality control, and forensic applications

More About this Item

Pipette tips sterilized by beta radiation maintain purity greater than those exposed to ethylene oxide, which is a gas absorbed by plastics.

Additional certifications for Biopur tips include DNA-free, DNase-free, RNase-free, PCR inhibitor-free, and ATP-free.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat