Eppendorf epTIPS Biopur and Sterile Pipette Tips
High standards of purity ideally suited for critical laboratory applications
  • Pyrogen-free (endotoxin-free) according to LAL test
  • Sterilized by beta radiation to SAL 10-6, avoiding contamination by ethylene oxide
  • Tested and certified by an independent laboratory
Pipette tips sterilized by beta radiation maintain purity greater than those exposed to ethylene oxide, which is a gas absorbed by plastics.
10,700.00 - ₹57,000.00INR / Case of 100
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Package Of
Availability Pricing
Eppendorf epTIPS Individually Wrapped Singles 0.1 to 20 uL 100/Pk 1888800 Mfr # 022491130
COMPARE
0.02 Individually Wrapped Singles; 100/Pk
₹13,200.00INR / Case of 100
Eppendorf epTIPS Individually Wrapped Singles 2 to 200 uL 100/Pk 1888802 Mfr # 022491148
COMPARE
0.2 Individually Wrapped Singles; 100/Pk
₹10,700.00INR / Case of 100
Eppendorf epTIPS Individually Wrapped Singles 50 to 1000 uL 100/Pk 1888804 Mfr # 022491156
COMPARE
1 Individually Wrapped Singles; 100/Pk
₹11,000.00INR / Case of 100
Eppendorf epTIPS Single Use Racks 2 to 200 uL 480/Pk 1888808 Mfr # 0030075234
COMPARE
0.2 Individually Wrapped Single Use Racks; 480/Pk
₹35,400.00INR / Case of 480
Eppendorf epTIPS Single Use Racks 50 to 1000 uL 480/Pk 1888812 Mfr # 0030075250
COMPARE
1 Individually Wrapped Single Use Racks; 480/Pk
₹36,500.00INR / Case of 480
Eppendorf epTIPS Sterile , Individually Wrapped Singles, 2 to 200 uL, 1000/CS 1888952 Mfr # 022492209
COMPARE
0.2 Individually Wrapped Singles; 1000/CS
₹57,000.00INR / Case of 1000
Eppendorf epTIPS Sterile , Individually Wrapped Singles, 50 to 1000 uL, 1000/CS 1888954 Mfr # 022492225
COMPARE
1 Individually Wrapped Singles; 1000/CS
₹57,000.00INR / Case of 1000
Eppendorf epTIPS Sterile , Single Use Racks, 20 to 300 uL, 960/CS 1888960 Mfr # 0030071573
COMPARE
0.3 Individually Wrapped Single Use Racks; 960/CS
₹36,200.00INR / Case of 960
Eppendorf epTIPS Sterile , Single Use Racks, 100 to 5000 uL, 120/CS 1888966 Mfr # 0030071620
COMPARE
5 Individually Wrapped Single Use Racks; 120/CS
₹39,600.00INR / Case of 120
Showing 1- 9 of 9