Film, 2 Mil, no adhesive, 12'' x 12''
Representative image only.
Item # 0827786
₹4,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)2
  • MaterialPolymide
  • Width (in)12
  • Length (in)12
  • Width (cm)30.5
More About this Item
Kapton® Type HN film is a tough, aromatic polyimide film, exhibiting an excellent balance of physical, chemical, and electrical properties over a wide temperature range, particularly at unusually high temperatures. It has been used successfully in applications at temperatures as low as -452°F (-269°C) and as high as 752°F (400°C). HN film can be laminated, metallized, punched, formed or adhesive coated.