Gardco EZ5 Series Viscosity Cup #5, 161 to 1401 Centistokes

Representative image only.
Gardco Mfr # EZ5 – Item # 0870040
An easy and economical way to measure viscosity
  • ±3% production tolerance

PRODUCT OPTIONS

₹60,100.00INR / Each

₹25,500.00INR / Each

for an additional
₹25,500.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Viscosity161 to 1401 centistokes
  • Sample Volume Needed (mL)46.5
  • Cup Number5
  • Nist-TraceableNo

Key Features

  • ±3% production tolerance

More About this Item

Easily measure viscosity using an EZ viscosity cup and a stopwatch. Simply fill the cup by immersing it in the test liquid. After withdrawing the cup, measure the time it takes until the first break in the liquid stream. Each cup is supplied with a conversion table to determine viscosity in centistokes (cSt) from the elapsed time.
Each stainless steel cup meets dimensions as specified in ASTM D 4212 standards.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat