EZ Series Viscosity Cups
An easy and economical way to measure viscosity
  • ±3% production tolerance
Easily measure viscosity using an EZ viscosity cup and a stopwatch. Simply fill the cup by immersing it in the test liquid. After withdrawing the cup, measure the time it takes until the first break in the liquid stream. Each cup is supplied with a conversion table to determine viscosity in centistokes (cSt) from the elapsed time.
Each stainless steel cup meets dimensions as specified in ASTM D 4212 standards.
58,700.00 - ₹130,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Cup Number Nist-Traceable Viscosity
Availability Pricing
Gardco EZ1 Series Viscosity Cup #1, 10 to 36 Centistokes 0870000 Mfr # EZ1
COMPARE
1 No 10 to 36 centistokes
₹60,100.00INR / Each
Gardco EZ1/C series viscosity cup #1 with certificate 0870002 Mfr # EZ1/C
COMPARE
1 Yes 10 to 36 centistokes
₹130,000.00INR / Each
Gardco EZ2 Series Viscosity Cup #2, 19 to 156 Centistokes 0870010 Mfr # EZ2
COMPARE
2 No 19 to 156 centistokes
₹60,100.00INR / Each
Gardco EZ2/C series viscosity cup #2 with certificate 0870012 Mfr # EZ2/C
COMPARE
2 Yes 19 to 156 centistokes
₹130,000.00INR / Each
Gardco EZ3 Series Viscosity Cup #3, 64 to 596 Centistokes 0870020 Mfr # EZ3
COMPARE
3 No 64 to 596 centistokes
₹60,100.00INR / Each
Gardco EZ3/C series viscosity cup #3 with certificate 0870022 Mfr # EZ3/C
COMPARE
3 Yes 64 to 596 centistokes
₹127,000.00INR / Each
Gardco EZ4 Series Viscosity Cup #4, 79 to 784 Centistokes 0870030 Mfr # EZ4
COMPARE
4 No 79 to 784 centistokes
₹58,700.00INR / Each
Gardco EZ4/C series viscosity cup #4 with certificate 0870032 Mfr # EZ4/C
COMPARE
4 Yes 79 to 784 centistokes
₹124,000.00INR / Each
Gardco EZ5 Series Viscosity Cup #5, 161 to 1401 Centistokes 0870040 Mfr # EZ5
COMPARE
5 No 161 to 1401 centistokes
₹60,100.00INR / Each
Gardco EZ5/C series viscosity cup #5 with certificate 0870042 Mfr # EZ5/C
COMPARE
5 Yes 161 to 1401 centistokes
₹127,000.00INR / Each
Showing 1- 10 of 10