Gardco VI-3321 Mini Ford Dip Viscosity Cup, Orifice #1, 12 to 40 Centistokes
Representative image only.
Gardco Mfr # VI-3321 – Item # 0870911
Feature handle for easy sample dipping – great for field testing
  • Compatible with ASTM D 1200
  • 1 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Sample Volume Needed (mL)67.5
  • Cup Number1
  • Warranty1 year
1 Year Warranty
Key Features
  • Compatible with ASTM D 1200
  • 1 year warranty
More About this Item
Take viscosity measurements of very thin paints, inks, solvents, adhesives, and coating materials of all kinds. Simply fill the cup by immersing it in the test liquid. After withdrawing the cup, measure the time it takes until the first break in the liquid stream. Each cup is supplied with a conversion table to determine viscosity in centistokes (cSt) from the elapsed time. Dip cups have an anodized aluminum handle for easy dipping and holding during timing. Replaceable orifice snaps in quickly and easily — no need to replace the whole cup.
Mini Ford Dip Viscosity Cups have half the capacity as the standard cups—perfect for smaller samples. Tolerance is ±4%.