Ford Dip Viscosity Cups
Feature handle for easy sample dipping – great for field testing
  • Compatible with ASTM D 1200
Take viscosity measurements of very thin paints, inks, solvents, adhesives, and coating materials of all kinds. Simply fill the cup by immersing it in the test liquid. After withdrawing the cup, measure the time it takes until the first break in the liquid stream. Each cup is supplied with a conversion table to determine viscosity in centistokes (cSt) from the elapsed time. Dip cups have an anodized aluminum handle for easy dipping and holding during timing. Replaceable orifice snaps in quickly and easily — no need to replace the whole cup.
80,200.00 - ₹147,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cup Number Sample Volume Needed (mL)
Availability Pricing
Gardco VI-3403 Standard Ford Dip Viscosity Cup, orifice #3 0870900 Mfr # VI-3403
COMPARE
3 135
₹140,000.00INR / Each
Gardco VI-3404 Standard Ford Dip Viscosity Cup, orifice #4 0870901 Mfr # VI-3404
COMPARE
4 135
₹147,000.00INR / Each
Gardco VI-3405 Standard Ford Dip Viscosity Cup, orifice #5 0870902 Mfr # VI-3405
COMPARE
5 135
Call For Price
Gardco VI-3320 Mini Ford Dip Viscosity Cup, Orifice #0, 2 to 14 Centistokes 0870910 Mfr # VI-3320
COMPARE
0 67.5
₹80,200.00INR / Each
Gardco VI-3321 Mini Ford Dip Viscosity Cup, Orifice #1, 12 to 40 Centistokes 0870911 Mfr # VI-3321
COMPARE
1 67.5
Call For Price
Gardco VI-3322 Mini Ford Dip Viscosity Cup, Orifice #2, 25 to 120 Centistokes 0870912 Mfr # VI-3322
COMPARE
2 67.5
₹84,300.00INR / Each
Gardco VI-3323 Mini Ford Dip Viscosity Cup, Orifice #3, 29 to 273 Centistokes 0870913 Mfr # VI-3323
COMPARE
3 67.5
Call For Price
Gardco VI-3324 Mini Ford Dip Viscosity Cup, Orifice #4, 53 to 441 Centistokes 0870914 Mfr # VI-3324
COMPARE
4 67.5
₹83,800.00INR / Each
Gardco VI-3325 Mini Ford Dip Viscosity Cup, Orifice #5, 215 to 1413 Centistokes 0870915 Mfr # VI-3325
COMPARE
5 67.5
Call For Price
Showing 1- 9 of 9