General Tools DCS200 Video Inspection Camera/Borescope, 12 mm dia Probe, 2.4" LCD
Representative image only.
General Tools Mfr # DCS200 – Item # 4861051
Ideal for inspection of duct work, piping, and wiring locations
  • Flexible camera-tipped probes
  • Adjustable LED lighting
  • Captures still and video images
  • 1 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 Year
1 Year Warranty
Includes
Probe, mirror, magnetic pickup, pickup hook, 9V battery, and custom hard plastic carrying case.
Key Features
  • Flexible camera-tipped probes
  • Adjustable LED lighting
  • Captures still and video images
  • 1 year warranty
More About this Item
These video borescope/inspection scopes are ideal for inspection of duct work, piping, wiring locations, and much more. Camera-tipped probes are flexible and strong. View findings instantaneously on the LCD monitor; some models will capture still and video images. Adjustable LED lighting on the probe tip provides illumination in dimly lit areas, plus allows you to lower the light intensity when viewing reflective objects to eliminate washout.
DCS200
* Economical units for a variety of applications
* IP67 rating on the probe and camera head (not the joints)
* 12 mm probe diameter for confined spaces