General Tools DCS800 Rugged High Performance VGA Recording Video Inspection System/Borescope
Representative image only.
General Tools Mfr # DCS800 – Item # 4861074
Ideal for inspection of duct work, piping, and wiring locations
  • Flexible camera-tipped probes
  • Adjustable LED lighting
  • Captures still and video images
  • 1 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Includes
90° mirrored probe, SD memory card (4 GB), USB and video cables, four AA batteries, adjustable hand strap, and custom hard plastic carring case.
Key Features
  • Flexible camera-tipped probes
  • Adjustable LED lighting
  • Captures still and video images
  • 1 year warranty
More About this Item
These video borescope/inspection scopes are ideal for inspection of duct work, piping, wiring locations, and much more. Camera-tipped probes are flexible and strong. View findings instantaneously on the LCD monitor; some models will capture still and video images. Adjustable LED lighting on the probe tip provides illumination in dimly lit areas, plus allows you to lower the light intensity when viewing reflective objects to eliminate washout.
DCS800
* Record VGA or QVGA resolution (640 x 480 pixel) video clips and still images
* Saved video and photo files incorporate real-time brightness, zoom, and mirroring adjustments
* Mirror live video either horizontal, vertical, or both
* 90° mirrored probe viewing tip
* Choice of 27 menu languages