General Tools P1618FS-49 Switchable Obedient Probe, 4.9 mm dia, 1 meter length
Representative image only.
General Tools Mfr # P1618FS-49 – Item # 4861067
Ideal for inspection of duct work, piping, and wiring locations
  • Flexible camera-tipped probes
  • Adjustable LED lighting
  • Captures still and video images
  • Compatible with DCS1600, DCS1700, DCS1800, and DCS800
  • 1 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Flexible camera-tipped probes
  • Adjustable LED lighting
  • Captures still and video images
  • Compatible with DCS1600, DCS1700, DCS1800, and DCS800
  • 1 year warranty
More About this Item
These video borescope/inspection scopes are ideal for inspection of duct work, piping, wiring locations, and much more. Camera-tipped probes are flexible and strong. View findings instantaneously on the LCD monitor; some models will capture still and video images. Adjustable LED lighting on the probe tip provides illumination in dimly lit areas, plus allows you to lower the light intensity when viewing reflective objects to eliminate washout.
The probe has two cameras that can be toggled to provide different views of the same target in a narrow space.