GF Signet 3-2760-1 DryLoc Preamp with 3/4" NPT threads, 15-ft cable
Representative image only.
GF Signet Mfr # 3-2760-1 – Item # 5656003
Customize your transmitter with add-on modules
  • Quickly form robust pH/ORP measurement assemblies
  • In-line integral mount and submersible installation versions
  • Get up and running quickly—auto configuration for pH and ORP operation
PRODUCT OPTIONS
₹126,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Thread Size NPT(M) (in)3/4
  • ConnectorDryLoc (ISO 7/1 R3/4)
  • Cable Length (m)4.5
Key Features
  • Quickly form robust pH/ORP measurement assemblies
  • In-line integral mount and submersible installation versions
  • Get up and running quickly—auto configuration for pH and ORP operation
More About this Item
These preamplifiers will allow any DryLoc pH/ORP electrodes to work with most +GF+ SIGNET instruments. These devices will self-configure for pH or ORP operation via automatic recognition of the electrode type. Electrodes with EasyCal allow simple push-button calibration and include an LED indicator for visual feedback. The DryLoc electrode connector forms a robust assembly for submersible and in-line installations. NEMA 4X junction boxes are integral parts of in-line versions and are available as accessories for submersible versions.