+GF+ Signet DryLoc pH and ORP Electrode Preamplifiers and Sensor Electronics
Customize your transmitter with add-on modules
  • Quickly form robust pH/ORP measurement assemblies
  • In-line integral mount and submersible installation versions
  • Get up and running quickly—auto configuration for pH and ORP operation
These preamplifiers will allow any DryLoc pH/ORP electrodes to work with most +GF+ SIGNET instruments. These devices will self-configure for pH or ORP operation via automatic recognition of the electrode type. Electrodes with EasyCal allow simple push-button calibration and include an LED indicator for visual feedback. The DryLoc electrode connector forms a robust assembly for submersible and in-line installations. NEMA 4X junction boxes are integral parts of in-line versions and are available as accessories for submersible versions.
85,200.00 - ₹126,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GF Signet 3-2760-1 DryLoc Preamp with 3/4" NPT threads, 15-ft cable 5656003 Mfr # 3-2760-1
COMPARE
DryLoc Preamp with 3/4" NPT threads, 15-ft cable
₹126,000.00INR / Each
GF Signet 3-2760-2 DryLoc Preamp with ISO threads, 15-ft cable 5656004 Mfr # 3-2760-2
COMPARE
DryLoc Preamp with ISO threads, 15-ft cable
₹85,200.00INR / Each