GVS Hygiene SwabCheck™ Swabs, 4 mL; 125/PK
Representative image only.
Mfr # 10498404 – Item # 7890944
About 1000 times more sensitive than the conventional ATP method at microorganism detection
  • Mess-free microorganism detection
  • Offers both qualitative and semi-quantitative hygiene control
  • Sterile packed and ready to use
  • For the detection of high levels of microorganisms
  • 25/PK
  • 1 year warranty
PRODUCT OPTIONS
₹212,000.00INR / Pkg of 125
OFTEN BOUGHT WITH…
Cole-Parmer® ThinTouch Nitrile Gloves, AQL 1.5
530.00 - ₹14,500.00INR / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeHygiene
  • Number of Tests Included125
  • DescriptionHygiene SwabCheck™ Swabs, 4 mL; 125/PK
  • Warranty1 year
1 Year WarrantySterile
Includes
Capped vials with media and a swab and a certificate of conformance (CoC) with item number, lot number, and expiration date.
Key Features
  • Mess-free microorganism detection
  • Offers both qualitative and semi-quantitative hygiene control
  • Sterile packed and ready to use
  • For the detection of high levels of microorganisms
  • 25/PK
  • 1 year warranty
More About this Item
Use the SwabCheck sterile cellulose swabs to quickly and easily check for the presence of bacteria on a surface. Simply wipe a surface with a swab, insert the swab into a tube containing a special medium with an indicator dye for a specific microorganism, then incubate. The super-sensitive SwabCheck medium changes color even when only a single bacterium is present. The quicker the color change happens, the higher the number of bacteria that are present. No color change after the maximum incubation period indicates the absence of the corresponding microorganism.
The hygiene SwabCheck swabs help monitor the levels of contamination at work areas and production lines to ensure they meet the requirements for cleanliness. A color change from purple to yellow indicates bacterial contamination; faster the change, higher the concentration. Each self-standing, round-bottomed plastic vial contains purple media with a cotton swab secured inside the screw cap. Store vials in temperatures between 2 and 8°C (35.6 and 46.4°F).