GVS SwabCheck™ Swab Kits
About 1000 times more sensitive than the conventional ATP method at microorganism detection
  • Mess-free microorganism detection
  • Offers both qualitative and semi-quantitative hygiene control
  • Sterile packed and ready to use
Use the SwabCheck sterile cellulose swabs to quickly and easily check for the presence of bacteria on a surface. Simply wipe a surface with a swab, insert the swab into a tube containing a special medium with an indicator dye for a specific microorganism, then incubate. The super-sensitive SwabCheck medium changes color even when only a single bacterium is present. The quicker the color change happens, the higher the number of bacteria that are present. No color change after the maximum incubation period indicates the absence of the corresponding microorganism.
91,100.00 - ₹217,000.00INR / Pkg of 30
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Tests Included Test Type Description
Availability Pricing
GVS Hygiene SwabCheck™ Swabs, 4 mL; 125/PK 7890944 Mfr # 10498404
COMPARE
125 Hygiene Hygiene SwabCheck™ Swabs, 4 mL; 125/PK
₹212,000.00INR / Pkg of 125
GVS <i>Escherichia coli</i> SwabCheck™ Swabs, 4 mL; 125/PK 7890945 Mfr # 10498402
COMPARE
125 Escherichia Escherichia coli SwabCheck™ Swabs, 4 mL; 125/PK
₹217,000.00INR / Pkg of 125
GVS Total Count Swab Kit for Enumeration of all Aerobic Bacteria on Surfaces; 30/PK 7890946 Mfr # 10498315
COMPARE
30 Total count Total Count Swab Kit for Enumeration of all Aerobic Bacteria on Surfaces; 30/PK
₹91,100.00INR / Pkg of 30
GVS Yeast and Mold Swab Kit; 30/PK 7890947 Mfr # 10498316
COMPARE
30 Yeast & Mold Yeast and Mold Swab Kit; 30/PK
₹91,100.00INR / Pkg of 30
GVS Coliform SwabCheck™ Swabs; 25/PK 7890948 Mfr # 10498406
COMPARE
25 Coliform Coliform SwabCheck™ Swabs; 25/PK
₹97,800.00INR / Pkg of 25
GVS Rapid Screening Hygiene SwabCheck™ Swabs; 25/PK 7890949 Mfr # 10498407
COMPARE
25 Hygiene Rapid Screening Hygiene SwabCheck™ Swabs; 25/PK
₹97,800.00INR / Pkg of 25
GVS <i>Listeria</i> SwabCheck™ Swabs; 25/PK 7890950 Mfr # 10498408
COMPARE
25 Listeria Listeria SwabCheck™ Swabs; 25/PK
₹97,800.00INR / Pkg of 25
Showing 1- 7 of 7