GVS Yeast and Mold Swab Kit; 30/PK
Representative image only.
Mfr # 10498316 – Item # 7890947
About 1000 times more sensitive than the conventional ATP method at microorganism detection
  • Mess-free microorganism detection
  • Offers both qualitative and semi-quantitative hygiene control
  • Sterile packed and ready to use
  • For the detection of yeast and mold on surfaces
  • 30/PK
  • 1 year warranty
PRODUCT OPTIONS
₹91,100.00INR / Pkg of 30
OFTEN BOUGHT WITH…
Cole-Parmer® ThinTouch Nitrile Gloves, AQL 1.5
530.00 - ₹14,500.00INR / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeYeast & Mold
  • Number of Tests Included30
  • DescriptionYeast and Mold Swab Kit; 30/PK
  • Warranty1 year
1 Year WarrantySterile
Includes
Capped vials with a swab, culture medium, nitrocellulose membrane device, and a certificate of conformance (CoC) with item number, lot number, and expiration date.
Key Features
  • Mess-free microorganism detection
  • Offers both qualitative and semi-quantitative hygiene control
  • Sterile packed and ready to use
  • For the detection of yeast and mold on surfaces
  • 30/PK
  • 1 year warranty
More About this Item
Use the SwabCheck sterile cellulose swabs to quickly and easily check for the presence of bacteria on a surface. Simply wipe a surface with a swab, insert the swab into a tube containing a special medium with an indicator dye for a specific microorganism, then incubate. The super-sensitive SwabCheck medium changes color even when only a single bacterium is present. The quicker the color change happens, the higher the number of bacteria that are present. No color change after the maximum incubation period indicates the absence of the corresponding microorganism.
Use this kit for the enumeration of yeast and mold on surfaces in accordance with Hazard Analysis and Critical Control Points (HACCP). Each self-standing, round-bottomed plastic vial contains green media with a cotton swab secured inside the screw cap. Obtain a quatitative result using the swabs, culture medium, and a membrane device included in the kit. Green opaque colonies against a yellow background are indicative of the growth of yeasts; mold colonies are green and filamentous. Store kits in temperatures between 2 and 8°C (35.6 and 46.4°F).