Humboldt H-5890 Accuflame Bunsen Burner, Adjustable with Threaded Needle Valve, Lp Gas
Representative image only.
Humboldt Mfr # H-5890 – Item # 3613002
  • 3 month warranty
PRODUCT OPTIONS
₹7,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Gas TypeLiquid Propane (LP)
  • BTU Output (per hour)3200
  • Inlet Tube OD (in)0.3125
  • Height (in)6 1/8
  • Warranty3 Months
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Adjustable Bunsen burners with threaded needle valve feature aluminum mixing tubes for thorough air/gas mixing; and tapered serrated gas inlet connections for easy tube mounting. Bases are plated die-cast zinc alloy. Shpg wt of all models is 0.5 lb (0.3 kg)