Straight Tube Bunsen Burners with Threaded Needle Valve
Adjustable Bunsen burners with threaded needle valve feature aluminum mixing tubes for thorough air/gas mixing; and tapered serrated gas inlet connections for easy tube mounting. Bases are plated die-cast zinc alloy. Shpg wt of all models is 0.5 lb (0.3 kg)
7,600.00 - ₹8,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
BTU Output (per hour) Gas Type Inlet Tube OD (in)
Availability Pricing
Humboldt H-5885 Accuflame Bunsen Burner, Adjustable with Threaded Needle Valve, Natural Gas 3613000 Mfr # H-5885
COMPARE
1200 Natural Gas 0.3125
₹8,100.00INR / Each
Humboldt H-5890 Accuflame Bunsen Burner, Adjustable with Threaded Needle Valve, Lp Gas 3613002 Mfr # H-5890
COMPARE
3200 Liquid Propane (LP) 0.3125
₹7,600.00INR / Each
Humboldt H-5880 Accuflame Bunsen Burner, Adjustable with Threaded Needle Valve, Artificial Gas 3613004 Mfr # H-5880
COMPARE
800 Artificial Gas 0.3125
₹7,700.00INR / Each