Idex Low-Pressure Y Connector Body, Natural PEEK, 0.060" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA

Representative image only.
Idex Mfr # P-514-01 – Item # 0202333
Combine, split, or intersect fluid streams throughout your system
  • Manufactured from inert polymers
  • Provide your own fittings to suit your application
  • For 1/8" OD tubing
  • 0.060" thru-hole diameter
  • 1 year warranty

PRODUCT OPTIONS

₹6,800.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(F) (port 1) (in)1/4-28 Flat Bottom
  • Compression Size / Tube Od (Port 2) (in)1/8
  • Compression Size / Tube Od (Port 2) (mm)3.2
  • ColorNatural
  • Max Operating Pressure (PSI)500
  • Orifice Dimensions (mm)1.5
  • DescriptionLow-Pressure Y Connector Body, Natural PEEK, 0.060" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
  • Warranty1 year
1 Year Warranty

Key Features

  • Manufactured from inert polymers
  • Provide your own fittings to suit your application
  • For 1/8" OD tubing
  • 0.060" thru-hole diameter
  • 1 year warranty

More About this Item

Use Y connectors in a variety of low-pressure situations throughout your fluidic system. Y connectors create less turbulence and solvent outgassing than Tee connectors, improving analytical results. Handy mounting holes make it a snap to mount connectors.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat