Idex Super Flangeless™ Ferrule, Natural PEEK Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex Mfr # P-360 – Item # 0202204
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Max pressure of 1500 psi (102 bar)
  • For 1/8" OD tubing
  • Natural PEEK/316 SS ring
  • 1/EA
  • 1 year warranty
PRODUCT OPTIONS
₹1,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPEEK/SS
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorNatural
  • Max Operating Pressure (PSI)1500
  • DescriptionSuper Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Max pressure of 1500 psi (102 bar)
  • For 1/8" OD tubing
  • Natural PEEK/316 SS ring
  • 1/EA
  • 1 year warranty
More About this Item
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.