Idex® Super Flangeless™ Ferrules, 1/4-28 Flat Bottom
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
960.00 - ₹17,700.00INR / Pkg of 10
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0201384 Mfr # P-350X
₹9,800.00INR / Pkg of 10
COMPARE
1/8 PEEK/SS Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK
Loading
₹9,800.00INR / Pkg of 10
Idex Super Flangeless™ Ferrule, Yellow ETFE/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0201385 Mfr # P-359X
₹8,900.00INR / Pkg of 10
COMPARE
1/8 ETFE/SS Super Flangeless™ Ferrule, Yellow ETFE/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK
Loading
₹8,900.00INR / Pkg of 10
Idex Super Flangeless™ Ferrule, Yellow ETFE; 1/EA 0202167 Mfr # P-258-01
₹1,300.00INR / Each
COMPARE
ETFE Super Flangeless™ Ferrule, Yellow ETFE; 1/EA
Loading
₹1,300.00INR / Each
Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202193 Mfr # P-350
₹1,000.00INR / Each
COMPARE
1/8 PEEK/SS Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Loading
₹1,000.00INR / Each
Idex Super Flangeless™ Ferrule, Natural PEEK Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202204 Mfr # P-360
₹1,900.00INR / Each
COMPARE
1/8 PEEK/SS Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Loading
₹1,900.00INR / Each
Idex Super Flangeless™ Ferrule, Natural PEEK Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0202205 Mfr # P-360X
₹17,700.00INR / Pkg of 10
COMPARE
1/8 PEEK/SS Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK
Loading
₹17,700.00INR / Pkg of 10
Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 2.5 mm OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202206 Mfr # P-366
₹1,100.00INR / Each
COMPARE
3/32 PEEK/SS Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 2.5 mm OD Tubing, 1/4-28 Flat Bottom; 1/EA
Loading
₹1,100.00INR / Each
Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 2.5 mm OD Tubing, 1/4-28 Flat Bottom; 10/PK 0202207 Mfr # P-366X
₹10,400.00INR / Pkg of 10
COMPARE
3/32 PEEK/SS Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 2.5 mm OD Tubing, 1/4-28 Flat Bottom; 10/PK
Loading
₹10,400.00INR / Pkg of 10
Idex Super Flangeless™ Ferrule, Yellow ETFE/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202315 Mfr # P-359
₹960.00INR / Each
COMPARE
1/8 ETFE/SS Super Flangeless™ Ferrule, Yellow ETFE/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Loading
₹960.00INR / Each
Idex Super Flangeless™ Ferrule Only, Yellow ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202316 Mfr # P-359-01
₹990.00INR / Each
COMPARE
1/8 ETFE Super Flangeless™ Ferrule Only, Yellow ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Loading
₹990.00INR / Each
Showing 1- 10 of 10