Labnet D1304-230V AccuBlock™ Dry Block Heater, Quad; 230 VAC

Representative image only.
Labnet Mfr # D1304-230V – Item # 5140123
Precise temperature regulation up to 150°C ensures accurate results
  • Single, dual, and four block configurations available
  • Dual digital displays
  • External probe available for better traceability
  • 3 year warranty

PRODUCT OPTIONS

Loading

₹211,000.00INR / Each
Loading

SPECS & DESCRIPTION

Specifications & Description

  • Length (")15 3/8
  • Number Of Blocks4
  • Width (InchShort)8 1/4
  • Height (InchShort)4 3/4
  • Uniformity (° C)0.2
  • Max Temperature (° F)302
  • Max Temperature (° C)150
  • Power (VAC)230
  • DescriptionLabnet D1304-230V AccuBlock™ Dry Block Heater, Quad; 230 VAC
  • Warranty3 year
3 Year WarrantyConformite EuropeenneCanadian Standards Association

More About this Item

AccuBlock Digital Dry baths are available in single, dual, and four block configurations. Ideal in a variety of applications including determining melting and boiling points, cell cultures, enzyme reactions and many more. Built of anodized, high grade non-porous aluminum, these units extremely durable and come with USB connectivity included.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat