Megger 1012-067 Temperature Probe, Type T, CAT III 600 V; for MTR105
Representative image only.
Megger Mfr # 1012-067 – Item # 2002661
Optimize performance of your Megger rotating machine tester
  • Compatible with Megger MTR105 Rotating Machine Tester (20026-56)
  • CAT III 600 V rated
  • 1 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeType T
  • Min Temperature (° F)4
  • Max Temperature (° F)392
  • Min Temperature (° C)-20
  • Max Temperature (° C)200
  • DescriptionTemperature Probe, Type T, CAT III 600 V; for MTR105
  • Warranty1 year
1 Year Warranty
Key Features
  • Compatible with Megger MTR105 Rotating Machine Tester (20026-56)
  • CAT III 600 V rated
  • 1 year warranty
More About this Item
Choose from the following line of Megger rotating machine tester accessories.