Megger Rotating Machine Tester Accessories

Optimize performance of your Megger rotating machine tester
  • Compatible with Megger MTR105 Rotating Machine Tester (20026-56)
Choose from the following line of Megger rotating machine tester accessories.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Megger 1012-064 Kelvin Probe Pins for MTR105; 4/pk 2002657 Mfr # 1012-064
COMPARE
Kelvin Probe Pins for MTR105; 4/pk
Call For Price
Megger 1012-065 Test Probe Set, Long Reach, CAT II 1000 V; for MTR105 2002658 Mfr # 1012-065
COMPARE
Test Probe Set, Long Reach, CAT II 1000 V; for MTR105
Call For Price
Megger 1012-066 Test Probe Set, Long Reach, CAT IV 600 V; for MTR105 2002659 Mfr # 1012-066
COMPARE
Test Probe Set, Long Reach, CAT IV 600 V; for MTR105
Call For Price
Megger 1012-069 Grabber Clip Lead Set, CAT III 100 V, CAT IV 600 V; for MTR105 2002660 Mfr # 1012-069
COMPARE
Grabber Clip Lead Set, CAT III 100 V, CAT IV 600 V; for MTR105
Call For Price
Megger 1012-067 Temperature Probe, Type T, CAT III 600 V; for MTR105 2002661 Mfr # 1012-067
COMPARE
Temperature Probe, Type T, CAT III 600 V; for MTR105
Call For Price
Megger 1012-068 Hook Strap Assembly Kit for MTR105 2002662 Mfr # 1012-068
COMPARE
Hook Strap Assembly Kit for MTR105
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat