Megger MPQ1000-B-KIT Power Quality Analyzer, Bronze Kit
Representative image only.
Megger Mfr # MPQ1000-B-KIT – Item # 2000065
Automatically recognizes current clamps and verifies proper connections
  • Measure RMS voltage and current, phase-to-phase voltage, power parameters, voltage and current dips and swells, and phase angles
  • Determine IEC unbalance, IEC flicker, and energy parameters (KWH, KVARH, and KVAH)
  • Capture event waveform, total harmonic distortion, total demand distortion, frequency, harmonics, and inter-harmonics
  • Power current clamps through the meter to avoid additional connections
  • Transfer data through USB, Ethernet, USB stick, or SD card
  • Large 32-GB memory card included with meter and meter packages
  • Free software—no license or dongle required
  • CATIV-600 V safety rating
  • 3 year warranty
PRODUCT OPTIONS
Call For Price
₹39,200.00INR / Each
for an additional
₹39,200.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max AC Voltage (Voltage)1000
  • AC Voltage Accuracy0.1% of reading
  • Max AC Current (amps)6000
  • AC Current Accuracy0.1% of reading + probe error
  • Max DC Voltage (Voltage)1000
  • Power Factor AccuracyAccuracy: ±0.02 + probe error
  • MemoryExternal SD card (32GB max)
  • Communication PortsUSB, Ethernet, USB stick, and SD card
  • Sample Rate256 samples per cycle RMS. 1 MHz transient sampling rate.
  • DisplayBacklit 11.5 cm x 8.5 cm color VGA
  • Warranty3 Years
3 Year WarrantyIP54 Rated
Includes
20026-38, Bronze Kit, adds four voltage lead plunger clips, hanging strap, and three 100 A current clamps.
Key Features
  • Measure RMS voltage and current, phase-to-phase voltage, power parameters, voltage and current dips and swells, and phase angles
  • Determine IEC unbalance, IEC flicker, and energy parameters (KWH, KVARH, and KVAH)
  • Capture event waveform, total harmonic distortion, total demand distortion, frequency, harmonics, and inter-harmonics
  • Power current clamps through the meter to avoid additional connections
  • Transfer data through USB, Ethernet, USB stick, or SD card
  • Large 32-GB memory card included with meter and meter packages
  • Free software—no license or dongle required
  • CATIV-600 V safety rating
  • 3 year warranty
More About this Item
Use the MPQ1000 to view RMS data, waveforms, demand data, phase angles, harmonics, unbalance, and flicker in real-time over three phases—on the color VGA display. Phase angle stability measurements make finding tricky phasing problems easy. Timed waveform capture allows for PQ event and periodic waveforms recording simultaneously.
This analyzer works in both scope and DMM modes. Choose from the meter to configure your own system, or select a package for additional savings.