Megger MPQ1000 Power Quality Analyzer
Automatically recognizes current clamps and verifies proper connections
  • Measure RMS voltage and current, phase-to-phase voltage, power parameters, voltage and current dips and swells, and phase angles
  • Determine IEC unbalance, IEC flicker, and energy parameters (KWH, KVARH, and KVAH)
  • Capture event waveform, total harmonic distortion, total demand distortion, frequency, harmonics, and inter-harmonics
  • Power current clamps through the meter to avoid additional connections
  • Transfer data through USB, Ethernet, USB stick, or SD card
  • Large 32-GB memory card included with meter and meter packages
  • Free software—no license or dongle required
  • CATIV-600 V safety rating
Use the MPQ1000 to view RMS data, waveforms, demand data, phase angles, harmonics, unbalance, and flicker in real-time over three phases—on the color VGA display. Phase angle stability measurements make finding tricky phasing problems easy. Timed waveform capture allows for PQ event and periodic waveforms recording simultaneously.
This analyzer works in both scope and DMM modes. Choose from the meter to configure your own system, or select a package for additional savings.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max AC Voltage (Voltage) Description
Availability Pricing
Megger MPQ1000-B-KIT Power Quality Analyzer, Bronze Kit 2000065 Mfr # MPQ1000-B-KIT
COMPARE
1000
Call For Price
Megger MPQ1000-S-KIT Power Quality Analyzer, Silver Kit 2000066 Mfr # MPQ1000-S-KIT
COMPARE
1000
Call For Price
Megger MPQ1000-G-KIT Power Quality Analyzer, Gold Kit 2000067 Mfr # MPQ1000-G-KIT
COMPARE
1000