Mettler Toledo Redox Ag ORP Electrode; S7
Representative image only.
Mettler Toledo Mfr # 51343205 – Item # 3590235
Precision ORP probes to monitor oxidation-reduction reactions
  • Cmbination ORP electrodes
  • Cable not included
PRODUCT OPTIONS
₹156,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsOxidation-reduction
  • Electrode TypeSilver
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference Electrolyte1M KNO3
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • ATC Element
  • ConnectorS7
  • Diameter (mm)12
  • Length (mm)120
Key Features
  • Cmbination ORP electrodes
  • Cable not included
More About this Item
The ARGENTHAL™ reference system is silver free—the junction won't become blocked by sulfides or proteins.
Select the appropriate sensor cable for your application and meter—BNC, waterproof BNC, DIN, Lemo OO, and wire-only cables available.